Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc27a3
Entrez Gene ID
26568
Gene Name
solute carrier family 27 (fatty acid transporter), member 3
Gene Aliases
Acsvl3, FATP3, Vlcs-3
Chromosome Location
Chr.3: 90385228 - 90390042 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011988.2 NP_036118.2 1 - 2 752 150
GenBank mRNA AF072758.1 - 1 - 2 456 150
AJ577573.1 - 1 - 2 752 150
AK076014.1 - 1 - 2 484 150
AK135190.1 - 1 - 2 729 150
BC060052.1 - 1 - 2 763 150
BC141134.1 - 1 - 2 717 150

View Full Product Details