Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc27a3
Entrez Gene ID
26568
Gene Name
solute carrier family 27 (fatty acid transporter), member 3
Gene Aliases
Acsvl3, FATP3, Vlcs-3
Chromosome Location
Chr.3: 90385228 - 90390042 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011988.2 NP_036118.2 9 - 10 1966 62
GenBank mRNA AF072758.1 - 9 - 10 1670 62
AJ577573.1 - 9 - 10 1966 62
AK037042.1 - 9 - 10 1726 62
AK076014.1 - 9 - 10 1698 62
AK131156.1 - 3 - 4 3229 62
AK135190.1 - 9 - 10 1943 62
BC060052.1 - 7 - 8 2191 62
BC141134.1 - 9 - 10 1931 62
CJ309971.1 - 1 - 2 102 62

View Full Product Details