Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zbbx
Entrez Gene ID
213234
Gene Name
zinc finger, B-box domain containing
Gene Aliases
4931432L23
Chromosome Location
Chr.3: 75037895 - 75165050 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172515.2 NP_766103.2 2 - 3 187 104
GenBank mRNA AK029899.1 - 2 - 3 196 104
AK133102.1 - 2 - 3 238 104
BC050789.1 - 2 - 3 187 104
BC100435.1 - 2 - 3 187 104

View Full Product Details