Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nid1
Entrez Gene ID
18073
Gene Name
nidogen 1
Gene Aliases
A630025O17, Nid, entactin, entactin-1, nidogen-1
Chromosome Location
Chr.13: 13437602 - 13512275 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010917.2 NP_035047.2 16 - 17 3323 98
GenBank mRNA AK084876.1 - 16 - 17 3262 98
AK144878.1 - 16 - 17 3288 98
AK166779.1 - 16 - 17 3259 98
BC052939.1 - 16 - 17 3318 98
BC131669.1 - 16 - 17 3234 98
X14194.1 - 16 - 17 3234 98
X14480.1 - 16 - 17 3244 98

View Full Product Details