Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Atp11a
Entrez Gene ID
50770
Gene Name
ATPase, class VI, type 11A
Gene Aliases
4930558F19Rik, AU040868, Atpc1h, Ih
Chromosome Location
Chr.8: 12757016 - 12868728 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001293667.1 NP_001280596.1 19 - 20 2546 111
NM_001293668.1 NP_001280597.1 19 - 20 2546 111
NM_015804.3 NP_056619.1 19 - 20 2546 111
NR_121603.1 19 - 20 2546 111
GenBank mRNA AF156551.1 - 19 - 20 2338 111
AK147497.1 - 19 - 20 2549 111
AK173088.1 - 19 - 20 2699 111
BC036322.1 - 4 - 5 607 111
BC138715.1 - 19 - 20 2265 111
BC145399.1 - 19 - 20 2266 111
BC145400.1 - 19 - 20 2266 111
CF167530.1 - 1 - 2 86 111

View Full Product Details