Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Chrna9
Entrez Gene ID
231252
Gene Name
cholinergic receptor, nicotinic, alpha polypeptide 9
Gene Aliases
2410015I05Rik, Acra9, EG666827, Gm8311
Chromosome Location
Chr.5: 65934764 - 65977560 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081104.1 NP_001074573.1 1 - 2 383 67
GenBank mRNA AK010496.1 - 1 - 2 382 67

View Full Product Details