The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081104.1 | NP_001074573.1 | 4 - 5 | 1219 | 63 |
GenBank mRNA | AK010496.1 | - | 4 - 5 | 1212 | 63 |
AY095483.1 | - | 1 - 2 | 389 | 63 |