Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Slc17a9
- Entrez Gene ID
-
228993
- Gene Name
- solute carrier family 17, member 9
- Gene Aliases
-
1700019H03Rik, BC019537, Vnut
- Chromosome Location
-
Chr.2: 180725213 - 180742280 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_183161.3 |
NP_898984.3 |
4 - 5 |
553 |
102 |
View Full Product Details