Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ghitm
Entrez Gene ID
66092
Gene Name
growth hormone inducible transmembrane protein
Gene Aliases
1010001P14Rik, C77840, MICS1, PTD010, Tmbim5
Chromosome Location
Chr.14: 37120444 - 37135327 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199122.1 NP_001186051.1 7 - 8 996 69
NM_078478.5 NP_510963.1 7 - 8 949 69
GenBank mRNA AF412297.1 - 7 - 8 893 69
AK005636.1 - 7 - 8 894 69
AK093887.1 - 7 - 8 925 69
AK134592.1 - 7 - 8 879 69
AK146433.1 - 7 - 8 856 69
AK146853.1 - 7 - 8 912 69
AK151488.1 - 7 - 8 926 69
AK158904.1 - 7 - 8 939 69
AK159695.1 - 7 - 8 923 69
AK159724.1 - 7 - 8 923 69
AK164122.1 - 7 - 8 894 69
AK167970.1 - 7 - 8 920 69
AK168022.1 - 7 - 8 926 69
AK168678.1 - 7 - 8 997 69
AK168800.1 - 7 - 8 905 69
AK189963.1 - 1 - 2 114 69
BC008622.1 - 7 - 8 927 69
BC010224.1 - 7 - 8 929 69

View Full Product Details