Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Actr8
Entrez Gene ID
56249
Gene Name
ARP8 actin-related protein 8
Gene Aliases
5730542K05Rik, ARP8
Chromosome Location
Chr.14: 29978330 - 29993221 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027493.3 NP_081769.2 2 - 3 419 89
GenBank mRNA AK017815.1 - 2 - 3 370 89
BC027281.1 - 2 - 3 325 89

View Full Product Details