The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033361.3 | NP_001028533.1 | 3 - 4 | 800 | 87 |
GenBank mRNA | AK136917.1 | - | 3 - 4 | 731 | 87 |
AK138339.1 | - | 3 - 4 | 801 | 87 | |
AY255586.1 | - | 2 - 3 | 521 | 87 |