The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026861.2 | NP_081137.2 | 6 - 7 | 700 | 71 |
GenBank mRNA | AK035384.1 | - | 3 - 4 | 275 | 71 |
AK049109.1 | - | 6 - 7 | 649 | 71 | |
AK158177.1 | - | 6 - 7 | 649 | 71 | |
AK159755.1 | - | 1 - 2 | 58 | 71 | |
BC024467.1 | - | 6 - 7 | 686 | 71 |