Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Rps6ka6
- Entrez Gene ID
-
67071
- Gene Name
- ribosomal protein S6 kinase polypeptide 6
- Gene Aliases
-
2610524K04Rik, RSK4, S6K-alpha-6
- Chromosome Location
-
Chr.X: 111387837 - 111538097 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_025949.3 |
NP_080225.2 |
9 - 10 |
1002 |
125 |
View Full Product Details