The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286647.1 | NP_001273576.1 | 4 - 5 | 500 | 105 |
NM_019717.3 | NP_062691.3 | 5 - 6 | 745 | 105 | |
NM_178050.4 | NP_835151.2 | 4 - 5 | 500 | 105 | |
GenBank mRNA | AK038697.1 | - | 4 - 5 | 428 | 105 |
AK135988.1 | - | 4 - 5 | 391 | 105 | |
BC055466.1 | - | 2 - 3 | 103 | 105 | |
BC060501.1 | - | 5 - 6 | 650 | 105 |