The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001161837.1 | NP_001155309.1 | 7 - 8 | 1060 | 131 |
NM_001161838.1 | NP_001155310.1 | 4 - 5 | 1017 | 131 | |
NM_001161839.1 | NP_001155311.1 | 5 - 6 | 708 | 131 | |
NM_001161840.1 | NP_001155312.1 | 5 - 6 | 900 | 131 | |
NM_011217.2 | NP_035347.1 | 8 - 9 | 1666 | 131 | |
GenBank mRNA | AF041866.1 | - | 5 - 6 | 825 | 131 |
AF129509.1 | - | 4 - 5 | 942 | 131 | |
AK028054.1 | - | 8 - 9 | 1667 | 131 | |
BC119230.1 | - | 8 - 9 | 1599 | 131 | |
BC120547.1 | - | 8 - 9 | 1599 | 131 | |
D31898.1 | - | 8 - 9 | 1629 | 131 | |
Z30313.1 | - | 7 - 8 | 1055 | 131 |