Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Kif4
Entrez Gene ID
16571
Gene Name
kinesin family member 4
Gene Aliases
AI323435, D330050K22Rik, Kns4
Chromosome Location
Chr.X: 100626065 - 100727271 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008446.2 NP_032472.2 25 - 26 3382 79
GenBank mRNA AK135430.1 - 25 - 26 2876 79
AK136346.1 - 6 - 7 623 79
AK171938.1 - 25 - 26 3294 79
BC050946.1 - 25 - 26 3320 79
BC054537.1 - 25 - 26 3304 79
D12646.1 - 25 - 26 3709 79

View Full Product Details