Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kif4
Entrez Gene ID
16571
Gene Name
kinesin family member 4
Gene Aliases
AI323435, D330050K22Rik, Kns4
Chromosome Location
Chr.X: 100626065 - 100727271 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008446.2 NP_032472.2 3 - 4 825 123
GenBank mRNA AK084837.1 - 3 - 4 802 123
AK135179.1 - 3 - 4 773 123
AK135430.1 - 3 - 4 319 123
AK171938.1 - 3 - 4 738 123
BC050946.1 - 3 - 4 763 123
BC054537.1 - 3 - 4 747 123
D12646.1 - 3 - 4 1152 123

View Full Product Details