The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136091.2 | NP_001129563.1 | 6 - 7 | 752 | 68 |
NM_001167784.1 | NP_001161256.1 | 6 - 7 | 1097 | 68 | |
NM_016921.3 | NP_058617.3 | 6 - 7 | 793 | 68 | |
GenBank mRNA | AB022322.1 | - | 6 - 7 | 659 | 68 |
AF218253.1 | - | 6 - 7 | 731 | 68 | |
AF218254.1 | - | 1 - 2 | 27 | 68 | |
AK155140.1 | - | 6 - 7 | 1098 | 68 | |
BC006761.1 | - | 6 - 7 | 793 | 68 | |
BC085234.1 | - | 6 - 7 | 653 | 68 |