The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144868.3 | NP_659117.2 | 27 - 28 | 5226 | 61 |
GenBank mRNA | AF237953.1 | - | 25 - 26 | 3243 | 61 |
AK144114.1 | - | 3 - 4 | 486 | 61 | |
AK220471.1 | - | 5 - 6 | 1124 | 61 | |
BC038056.1 | - | 22 - 23 | 3334 | 61 | |
BC054751.1 | - | 27 - 28 | 4505 | 61 | |
BC055289.1 | - | 28 - 29 | 5029 | 61 | |
BC096538.1 | - | 27 - 28 | 5226 | 61 |