Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tmem89
Entrez Gene ID
69384
Gene Name
transmembrane protein 89
Gene Aliases
1700024B07Rik
Chromosome Location
Chr.9: 108914619 - 108915563 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027066.1 NP_081342.1 1 - 2 339 74
GenBank mRNA AK006292.1 - 1 - 2 341 74
BY706371.1 - 1 - 2 341 74

View Full Product Details