The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001161362.3 | NP_001154834.1 | 9 - 10 | 3447 | 107 |
NM_172144.3 | NP_742156.2 | 8 - 9 | 1287 | 107 | |
GenBank mRNA | AK035077.1 | - | 8 - 9 | 1067 | 107 |
AK076163.1 | - | 8 - 9 | 2499 | 107 | |
AK132175.1 | - | 8 - 9 | 1288 | 107 | |
BC042841.1 | - | 8 - 9 | 2408 | 107 | |
BC042842.1 | - | 8 - 9 | 2881 | 107 | |
BC151170.1 | - | 8 - 9 | 3274 | 107 | |
BC151171.1 | - | 8 - 9 | 3269 | 107 |