Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Las1l
Entrez Gene ID
76130
Gene Name
LAS1-like (S. cerevisiae)
Gene Aliases
1810030A06Rik, 5830482G23Rik
Chromosome Location
Chr.X: 95920045 - 95956974 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152822.3 NP_690035.2 3 - 4 516 115
GenBank mRNA AK018059.1 - 3 - 4 446 115
BC004735.1 - 2 - 3 108 115
BC032218.1 - 3 - 4 439 115
BC141155.1 - 3 - 4 453 115
BQ573371.1 - 2 - 3 96 115

View Full Product Details