Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ptprr
Entrez Gene ID
19279
Gene Name
protein tyrosine phosphatase, receptor type, R
Gene Aliases
Gmcp1, PTP-SL, PTPBR7, RPTPRR, mPTP213
Chromosome Location
Chr.10: 116018200 - 116274932 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161840.1 NP_001155312.1 1 - 2 246 73
GenBank mRNA DV657487.1 - 1 - 2 246 73

View Full Product Details