Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mdm2
Entrez Gene ID
17246
Gene Name
transformed mouse 3T3 cell double minute 2
Gene Aliases
1700007J15Rik, AA415488, Mdm-2
Chromosome Location
Chr.10: 117688875 - 117710770 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001288586.1 NP_001275515.1 10 - 11 1095 123
NM_010786.4 NP_034916.1 11 - 12 1180 123
GenBank mRNA AK004719.1 - 11 - 12 1139 123
AK004881.1 - 9 - 10 2322 123
AK088638.1 - 11 - 12 1110 123
AK150530.1 - 9 - 10 871 123
AK152685.1 - 9 - 10 871 123
AK160069.1 - 10 - 11 971 123
AK168690.1 - 11 - 12 1163 123
BC050902.1 - 11 - 12 1096 123
BC092270.1 - 11 - 12 1127 123
U47934.1 - 9 - 10 886 123
X58876.1 - 11 - 12 1087 123

View Full Product Details