Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ttc39d
Entrez Gene ID
67737
Gene Name
tetratricopeptide repeat domain 39D
Gene Aliases
4930560E09Rik
Chromosome Location
Chr.17: 80215914 - 80217936 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026351.2 NP_080627.2 1 - 1 511 87
GenBank mRNA AK016184.1 - 1 - 1 94 87
BC118946.1 - 1 - 1 511 87
BC118947.1 - 1 - 1 511 87

View Full Product Details