Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Prkar2a
Entrez Gene ID
19087
Gene Name
protein kinase, cAMP dependent regulatory, type II alpha
Gene Aliases
1110061A24Rik, AI317181, AI836829, RII(alpha)
Chromosome Location
Chr.9: 108692143 - 108749512 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008924.2 NP_032950.1 7 - 8 1108 139
GenBank mRNA AF533977.1 - 7 - 8 1108 139
AK132728.1 - 7 - 8 1047 139
AK132824.1 - 7 - 8 1039 139
BC075623.1 - 7 - 8 1079 139
BC080276.1 - 7 - 8 1102 139
J02935.1 - 7 - 8 791 139

View Full Product Details