Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wdr43
Entrez Gene ID
72515
Gene Name
WD repeat domain 43
Gene Aliases
2610318G08Rik, AU020887, AV024208, Wrd43, mKIAA0007
Chromosome Location
Chr.17: 71615900 - 71659031 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175639.1 NP_783570.1 6 - 7 890 77
GenBank mRNA AK012043.1 - 6 - 7 875 77
AK032907.1 - 6 - 7 864 77
AK129031.1 - 6 - 7 808 77
AK152321.1 - 6 - 7 880 77
BC079913.1 - 5 - 6 591 77
BC117724.2 - 7 - 8 1095 77

View Full Product Details