Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Apoo
- Entrez Gene ID
-
68316
- Gene Name
- apolipoprotein O
- Gene Aliases
-
0610008C08Rik, 1110019O03Rik
- Chromosome Location
-
Chr.X: 94367011 - 94417093 on Build GRCm38
- Gene Symbol
- Apoo-ps
- Entrez Gene ID
-
621156
- Gene Name
- apolipoprotein O, pseudogene
- Gene Aliases
-
Apoo, EG621156
- Chromosome Location
-
Chr.13: 107414163 - 107414767 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_004438.1 |
|
1 - 1 |
410 |
103 |
GenBank mRNA |
BI716015.1 |
- |
1 - 1 |
379 |
103 |
CV674789.1 |
- |
1 - 1 |
395 |
103 |
View Full Product Details