The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026576.3 | NP_080852.2 | 2 - 3 | 436 | 121 |
GenBank mRNA | AK019965.1 | - | 2 - 3 | 437 | 121 |
BC028916.1 | - | 2 - 3 | 431 | 121 |