Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ift52
Entrez Gene ID
245866
Gene Name
intraflagellar transport 52
Gene Aliases
BC037708, NGD5
Chromosome Location
Chr.2: 163017298 - 163046155 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK144188.1 - 1 - 2 165 63

View Full Product Details