The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153195.2 | NP_694875.2 | 1 - 2 | 221 | 70 |
GenBank mRNA | AK147797.1 | - | 1 - 2 | 173 | 70 |
AK150626.1 | - | 1 - 2 | 179 | 70 | |
AK151025.1 | - | 1 - 2 | 179 | 70 | |
AK152513.1 | - | 1 - 2 | 179 | 70 | |
BC032153.1 | - | 1 - 2 | 163 | 70 | |
CD563204.1 | - | 1 - 2 | 221 | 70 |