Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Crispld2
Entrez Gene ID
78892
Gene Name
cysteine-rich secretory protein LCCL domain containing 2
Gene Aliases
1810049K24Rik, Lcrisp2, Lgl1, coffeecrisp
Chromosome Location
Chr.8: 119992443 - 120052793 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030209.3 NP_084485.1 9 - 10 1183 74
GenBank mRNA AK019034.1 - 9 - 10 1179 74
AK033858.1 - 9 - 10 1147 74
AK132171.1 - 9 - 10 1126 74
AK149909.1 - 9 - 10 1140 74
AK149925.1 - 9 - 10 1140 74
AK150180.1 - 9 - 10 1140 74
AK154308.1 - 9 - 10 1183 74
BC090642.1 - 9 - 10 1177 74

View Full Product Details