The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026648.4 | NP_080924.1 | 11 - 12 | 1942 | 93 |
GenBank mRNA | AK006656.1 | - | 4 - 5 | 601 | 93 |
AK019633.1 | - | 11 - 12 | 1942 | 93 | |
BC050751.1 | - | 11 - 12 | 1861 | 93 |