Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dnaaf1
Entrez Gene ID
68270
Gene Name
dynein, axonemal assembly factor 1
Gene Aliases
4930457P18Rik, Lrrc50
Chromosome Location
Chr.8: 119575235 - 119598454 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026648.4 NP_080924.1 3 - 4 465 62
GenBank mRNA AK019633.1 - 3 - 4 465 62
BC050751.1 - 3 - 4 384 62

View Full Product Details