The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001195090.1 | NP_001182019.1 | 4 - 5 | 630 | 159 |
NM_181856.2 | NP_862904.1 | 4 - 5 | 630 | 159 | |
GenBank mRNA | AK156764.1 | - | 4 - 5 | 591 | 159 |
AY236501.1 | - | 4 - 5 | 542 | 159 | |
AY263160.1 | - | 3 - 4 | 518 | 159 | |
BC138254.1 | - | 3 - 4 | 506 | 159 |