Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Olfr22-ps1
Entrez Gene ID
258209
Gene Name
olfactory receptor 22, pseudogene 1
Gene Aliases
MOR135-18, MTPCR35, Olfr22
Chromosome Location
Chr.11: 73954692 - 73955625 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA X89687.1 - 1 - 1 242 95

View Full Product Details