The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153145.4 | NP_694785.3 | 25 - 26 | 3509 | 103 |
GenBank mRNA | AB255881.1 | - | 24 - 25 | 3428 | 103 |
AB255882.1 | - | 25 - 26 | 3516 | 103 | |
AF498360.1 | - | 24 - 25 | 3437 | 103 | |
AK031843.1 | - | 6 - 7 | 740 | 103 | |
AY732492.1 | - | 24 - 25 | 3437 | 103 | |
BC026496.1 | - | 5 - 6 | 634 | 103 | |
BC060032.1 | - | 7 - 8 | 1003 | 103 |