Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rap1gap2
Entrez Gene ID
380711
Gene Name
RAP1 GTPase activating protein 2
Gene Aliases
AU067654, Garnl4, Gm1561, mKIAA1039
Chromosome Location
Chr.11: 74383483 - 74610910 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015046.2 NP_001015046.1 22 - 23 2268 94
GenBank mRNA AK038834.1 - 12 - 13 1331 94
AK122424.1 - 8 - 9 1061 94
AK147290.1 - 23 - 24 2245 94
BC107388.2 - 13 - 14 1474 94

View Full Product Details