The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001128086.2 | NP_001121558.2 | 19 - 20 | 3211 | 111 |
NM_001177397.1 | NP_001170868.1 | 5 - 6 | 657 | 111 | |
GenBank mRNA | AK032061.1 | - | 3 - 4 | 313 | 111 |
AK034154.1 | - | 6 - 7 | 677 | 111 | |
AK038609.1 | - | 3 - 4 | 541 | 111 | |
BC098373.1 | - | 5 - 6 | 657 | 111 |