Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gaa
Entrez Gene ID
14387
Gene Name
glucosidase, alpha, acid
Gene Aliases
E430018M07Rik
Chromosome Location
Chr.11: 119267967 - 119285698 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001159324.1 NP_001152796.1 11 - 12 1803 69
NM_008064.3 NP_032090.3 11 - 12 1893 69
GenBank mRNA AK052211.1 - 11 - 12 1874 69
AK088481.1 - 11 - 12 1869 69
AK139333.1 - 11 - 12 1841 69
AK140042.1 - 11 - 12 1861 69
AK141546.1 - 11 - 12 1876 69
AK145631.1 - 10 - 11 1309 69
AK146538.1 - 11 - 12 1797 69
AK150970.1 - 11 - 12 1851 69
BC010210.1 - 11 - 12 1857 69

View Full Product Details