Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zmynd8
Entrez Gene ID
228880
Gene Name
zinc finger, MYND-type containing 8
Gene Aliases
1110013E22Rik, 2010005I16Rik, 3632413B07Rik, AI316811, AL024039, Prkcbp1, RACK7
Chromosome Location
Chr.2: 165784152 - 165898984 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001252584.2 NP_001239513.1 12 - 13 1769 61
NM_001252585.1 NP_001239514.1 11 - 12 1661 61
NM_001281926.1 NP_001268855.1 12 - 13 1668 61
NM_001291158.1 NP_001278087.1 13 - 14 3778 61
NM_027230.5 NP_081506.3 13 - 14 1856 61
GenBank mRNA AB093284.2 - 13 - 14 3770 61
AK132427.1 - 11 - 12 1663 61
AK147620.1 - 12 - 13 1669 61
AK155864.1 - 12 - 13 1574 61
BC041797.1 - 2 - 3 584 61
BC048186.1 - 13 - 14 1663 61

View Full Product Details