Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ces5a
Entrez Gene ID
67935
Gene Name
carboxylesterase 5A
Gene Aliases
1700081L16Rik, 1700122C07Rik, BB081581, Ces7, Gm503, cauxin
Chromosome Location
Chr.8: 93499063 - 93535881 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003951.2 NP_001003951.1 7 - 8 913 76
GenBank mRNA AB186393.1 - 7 - 8 913 76

View Full Product Details