The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001029993.1 | NP_001025164.1 | 11 - 12 | 2057 | 103 |
NM_001029994.1 | NP_001025165.1 | 10 - 11 | 1985 | 103 | |
GenBank mRNA | AK031413.1 | - | 11 - 12 | 2007 | 103 |
AK131189.1 | - | 8 - 9 | 1129 | 103 | |
AK159881.1 | - | 11 - 12 | 1993 | 103 | |
AK165989.1 | - | 9 - 10 | 1339 | 103 | |
BC030495.1 | - | 10 - 11 | 1965 | 103 | |
BC094330.1 | - | 8 - 9 | 1138 | 103 |