Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fanca
Entrez Gene ID
14087
Gene Name
Fanconi anemia, complementation group A
Gene Aliases
AW208693, FACA
Chromosome Location
Chr.8: 123268244 - 123318651 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016925.3 NP_058621.2 2 - 3 213 90
GenBank mRNA AF208116.1 - 2 - 3 215 90
AF208118.1 - 2 - 3 215 90
AF208122.1 - 2 - 3 199 90
AF208123.1 - 2 - 3 180 90
AK082847.1 - 2 - 3 198 90
AK084990.1 - 2 - 3 198 90
AK166112.1 - 2 - 3 198 90
BC150705.1 - 2 - 3 211 90
CF537383.1 - 2 - 3 130 90

View Full Product Details