The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133973.2 | NP_598734.1 | 18 - 19 | 2250 | 73 |
GenBank mRNA | AK156466.1 | - | 18 - 19 | 2242 | 73 |
AK172465.1 | - | 18 - 19 | 2241 | 73 | |
AY223866.1 | - | 18 - 19 | 2277 | 73 | |
BC023120.1 | - | 18 - 19 | 2227 | 73 |