Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lin7a
Entrez Gene ID
108030
Gene Name
lin-7 homolog A (C. elegans)
Gene Aliases
AI848705, LIN-7A, MALS-1, TIP-33, Veli, Veli1
Chromosome Location
Chr.10: 107271798 - 107425146 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039354.1 NP_001034443.1 3 - 4 298 70
GenBank mRNA BC029721.1 - 3 - 4 204 70
BC105965.1 - 2 - 3 151 70
BG288381.1 - 3 - 4 236 70
BU923246.1 - 3 - 4 265 70
BY123635.1 - 3 - 4 298 70

View Full Product Details