The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001177616.1 | NP_001171087.1 | 7 - 8 | 862 | 90 |
NM_001177617.1 | NP_001171088.1 | 7 - 8 | 1029 | 90 | |
NM_001177618.1 | NP_001171089.1 | 6 - 7 | 560 | 90 | |
NM_001177619.1 | NP_001171090.1 | 7 - 8 | 1227 | 90 | |
NM_001177623.1 | NP_001171094.1 | 7 - 8 | 791 | 90 | |
NM_033264.2 | NP_150289.1 | 6 - 7 | 695 | 90 | |
GenBank mRNA | AF324451.1 | - | 6 - 7 | 655 | 90 |
AK029467.1 | - | 7 - 8 | 1228 | 90 | |
AK032417.1 | - | 7 - 8 | 788 | 90 | |
AK032543.1 | - | 5 - 6 | 475 | 90 | |
AK034376.1 | - | 5 - 6 | 408 | 90 | |
AK045507.1 | - | 7 - 8 | 1229 | 90 | |
AK050613.1 | - | 7 - 8 | 792 | 90 | |
AK137974.1 | - | 7 - 8 | 1030 | 90 | |
AK153892.1 | - | 8 - 9 | 1510 | 90 | |
BC053001.1 | - | 5 - 6 | 526 | 90 | |
BC098368.1 | - | 7 - 8 | 809 | 90 | |
BC128376.1 | - | 6 - 7 | 576 | 90 |