The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001141975.1 | NP_001135447.1 | 1 - 2 | 139 | 102 |
NM_001141976.1 | NP_001135448.1 | 1 - 2 | 139 | 102 | |
NM_001141978.1 | NP_001135450.1 | 1 - 2 | 139 | 102 | |
GenBank mRNA | AK090101.1 | - | 1 - 2 | 93 | 102 |
AK153957.1 | - | 1 - 2 | 74 | 102 | |
BU148033.1 | - | 1 - 2 | 41 | 102 | |
CJ163921.1 | - | 1 - 2 | 141 | 102 |