Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Atp2a3
Entrez Gene ID
53313
Gene Name
ATPase, Ca++ transporting, ubiquitous
Gene Aliases
SERCA3b, Serca3
Chromosome Location
Chr.11: 72961169 - 72993043 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163336.1 NP_001156808.1 16 - 17 2715 76
NM_001163337.1 NP_001156809.1 16 - 17 2715 76
NM_016745.3 NP_058025.2 16 - 17 2715 76
GenBank mRNA AK089487.1 - 16 - 17 2715 76
AK152999.1 - 3 - 4 587 76
AK155904.1 - 16 - 17 2716 76
BC017639.1 - 16 - 17 2629 76
BC026147.1 - 16 - 17 2661 76
U49393.1 - 16 - 17 2576 76
U49394.1 - 16 - 17 2576 76

View Full Product Details