Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Necab3
- Entrez Gene ID
-
56846
- Gene Name
- N-terminal EF-hand calcium binding protein 3
- Gene Aliases
-
2900010M17Rik, AI853434, Apba2bp, Nip1, XB51
- Chromosome Location
-
Chr.2: 154544399 - 154559406 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK013520.1 |
- |
8 - 9 |
715 |
74 |
View Full Product Details